Learn More
DWK Life Sciences Kimble™ Kontes™ Chromaflex Chromatography Fittings : Adapter with 1/2-20 to 1/4-28 Thread
Description
Converts 1/2-20 threads on CHROMAFLEX™ column end fittings to 1/4-28 threads.
- CTFE Threaded Adapter with 1/2-20 to 1/4-28 Thread
- Converts 1/2-20 threads on CHROMAFLEX™ column end fittings to 1/4-28 threads
- Case of 2
Specifications
Specifications
| Type | Adapter |
| For Use With (Application) | Chromatography |
| Packaging Quantity | 1 |
| Includes | Kimble Kontes Hi-Vac PTFE plug |
| Material | Glass |