Learn More
DWK Life Sciences Kimble™ Kontes™ Chromatography Column ETFE Ferrule for 1/16 in. Tubing
Catalog No. K4208220116
Change view
Click to view available options
Material:
ETFE
Catalog No. Material
K4208220116 ETFE
1 options
Catalog No. K4208220116 Supplier DWK Life Sciences Supplier No. 4208220116
Tubing nuts and ferrules are used to make connections between the tubing and the end fittings of CHROMAFLEX™, FLEX-COLUMN™ and DISPOSAFLEX™ columns.
  • EFTE Flangeless Ferrule for 1/16 in. OD Tubing
Specifications
For Use With (Equipment) 0.06 in. O.D. Tubing and Chromaflex Column Chromatography Fitting
Material ETFE
Type Ferrule
Quantity Case of 1