Learn More
DWK Life Sciences Kimble™ Kontes™ 105° Extended Bent Distillation Adapter with Vacuum Take-Off and Inner Hose Connection
Extended 105 degree bent distillation adapter with vacuum take-off and outer hose connection.
Supplier:  DWK Life Sciences 2764050000
Catalog No. K276405-0000
Only null left
Explore available promotions
Description
Description
  • Barbed hose connection
  • Standard Taper inner joint with drip tip at the bottom and Standard Taper outer joint at the top
  • Joints are the same size 14/20
  • Fits tubing ID 1/4 IN
  • Overall height 100mm
  • Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements
Specifications
Specifications
USP Type I, ASTM E438, Type I, Class A
6.35mm
100mm
Borosilicate Glass
14/20
0.25 in.
3.93 in.
Vacuum take-off, a hose connector and a drip tip
Distillation Bent Adapter
Product Suggestions
Product Suggestions