missing translation for 'onlineSavingsMsg'
Learn More

Kojic Acid 98.0+%, TCI America™

Catalog No. p-7131740
Change view
Click to view available options
Quantity:
25 g
500 g
Catalog No. Quantity
K001025G 25 g
K0010500G 500 g
2 options

Catalog No. K001025G

Supplier: TCI America K001025G

Only null left

Chemical Identifiers

CAS 501-30-4
Molecular Formula C6H6O4
Molecular Weight (g/mol) 142.11
MDL Number MFCD00006580
InChI Key BEJNERDRQOWKJM-UHFFFAOYSA-N
Synonym kojic acid, 5-hydroxy-2-hydroxymethyl-4h-pyran-4-one, 5-hydroxy-2-hydroxymethyl-4-pyrone, 4h-pyran-4-one, 5-hydroxy-2-hydroxymethyl, 5-hydroxy-2-hydroxymethyl pyran-4-one, acido kojico, 2-hydroxymethyl-5-hydroxy-gamma-pyrone, 2-hydroxymethyl-5-hydroxy-4h-pyran-4-one, unii-6k23f1tt52
PubChem CID 3840
ChEBI CHEBI:43572
IUPAC Name 5-hydroxy-2-(hydroxymethyl)-4H-pyran-4-one
SMILES OCC1=CC(=O)C(O)=CO1

Specifications

Melting Point 154°C
Color White-Yellow
Quantity 25 g
Formula Weight 142.11
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Kojic Acid
RTECSNumber UQ0875000
TSCA No