missing translation for 'onlineSavingsMsg'
Learn More

Kojic Acid, 99%, Spectrum™ Chemical

Catalog No. 18611167
Change view
Click to view available options
Quantity:
25 g
100 g
1 kg
5 kg
25 kg
50 kg
Packaging:
Amber Glass Bottle
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18611167 50 kg Amber Glass Bottle
18611163 25 g Amber Glass Bottle
18611162 100 g Amber Glass Bottle
18611161 1 kg Amber Glass Bottle
18611166 5 kg Amber Glass Bottle
18611164 25 kg Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 18611167 Supplier Spectrum Chemical Mfg Cor Supplier No. K106250KGBL

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 501-30-4
Molecular Formula C6H6O4
Molecular Weight (g/mol) 142.11
InChI Key BEJNERDRQOWKJM-UHFFFAOYSA-N
IUPAC Name 5-hydroxy-2-(hydroxymethyl)-4H-pyran-4-one
SMILES OCC1=CC(=O)C(O)=CO1

Specifications

CAS Max % 1
Loss on Drying 0.006
Packaging Amber Glass Bottle
Quantity 50 kg
Percent Purity 0.99
Grade Ungraded
Physical Form Crystalline Powder or Lumps
Chemical Name or Material Kojic Acid

CAUTION: For manufacturing, processing or repacking. Read and understand the label and Safety Data Sheet (SDS) prior to use.