Learn More
Koehler™ Instrument Petroleum Vapor Pressure Bomb
Supplier: Koehler™ Instrument K11500
Description
For ASTM D 323-99a
Full-size bomb for immersion sampling and testing of gasoline, volatile crude oil, and other petroleum products (except LPG) having vapor pressures of up to 26psi (180kPa). 4:1 ratio between air and gasoline chambers. All stainless steel.Supplied with O-ring-type seal for tight closure and 1/4-18 NPT threaded gauge connection. Order Vapor-Pressure Bomb Gauge separately.
Specifications
| Reid Vapor Pressure Cylinder | |
| Threaded 0.25 in. gauge coupling and 2 O-ring seals, upper chamber and lower chamber | |
| ASTM D323, D1267 |
| Stainless Steel | |
| Polished | |
| Immersion sampling and testing of gasoline, volatile crude oil and other petroleum products (except lpg) |