Learn More
Koehler™ Instrument Smoke Point Lamp and Accessories
Specifications
Specifications
| Product Type | Interchangeable Candle |
| Material | Plated Brass |
| Includes | Wick tube and air vent |
| For Use With (Application) | Burns fuel samples under controlled conditions for smoke point determinations of aviation turbine fuels and similar products |
| For Use With (Equipment) | Smoke point lamp |