Learn More
Koehler™ Instrument Rust Preventing Characteristics Oil Bath Test Specimens
Catalog No. KLK30110
Change view
Click to view available options
Includes:
w/out Holder
Catalog No. Includes
KLK30110 w/out Holder
1 options
Catalog No. KLK30110 Supplier Koehler™ Instrument Supplier No. K30110
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications
Product Type Test Specimen
Material Steel
Includes w/out Holder
Certifications/Compliance ASTM D665, D3603, NACE TM-01-72
For Use With (Application) Determine anti-rust property in oil bath apparatus
For Use With (Equipment) Oil bath apparatus