Learn More
Koehler™ Instrument LPG Copper Strip Corrosion Test Cylinder and Accessories
Catalog No. KLK40000
Change view
Click to view available options
Type:
Copper Strip
LPG Copper Strip Corrosion Test Cylinder
Quantity:
Each
One unit per pack
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type Quantity
KLK40000 LPG Copper Strip Corrosion Test Cylinder One unit per pack
KLK40200 Copper Strip Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. KLK40000 Supplier Koehler™ Instrument Supplier No. K40000
Specifications
Type LPG Copper Strip Corrosion Test Cylinder
Quantity One unit per pack
Includes 0.25 in. Needle valves for purging and admitting LPG samples, dip tube with hook, knurled, threaded cap with O-ring gasket hand tightens to a positive seal
Certifications/Compliance ASTM D1838, GPA 2140, ISO 6251, CE
Material Stainless Steel