missing translation for 'onlineSavingsMsg'
Learn More

Mopec Knife, Standard Grade Disposable, 10 pack

Catalog No. 22444258
Change view
Click to view available options
Length (English) Blade:
4 in.
6 in.
8 in.
10 in.
12 in.
Length (English):
8.5 in.
10.5 in.
12.5 in.
14.5 in.
16.5 in.
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Length (English) Blade Length (English)
22444258 4 in. 8.5 in.
AH025 6 in. 10.5 in.
AH026 8 in. 12.5 in.
AH027 10 in. 14.5 in.
AH028 12 in. 16.5 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 22444258 Supplier Mopec Supplier No. AH024

Disposable Mopec autopsy knives may be sharpened for a longer lasting edge. Non-slip handle ensures accurate dissection.

  • Disposable
  • May be sharpened for a longer lasting edge.
  • Non-slip handle ensures accurate dissection.

Specifications

Blade Material Stainless Steel
Product Type Standard Dissecting Knife
For Use With (Application) Dissection Labs
Handle Type Non-slip
Length (English) Blade 4 in.
Length (Metric) Blade 102 mm
Length (English) 8.5 in.
Disposable Single-use
Quantity 10/Pk.