Learn More
Mopec Knee Space Panel, Standing Height, Adjustable
Description
Construction
- (1) Standing height knee space panel, adjustable, for use with apron rail.
Materials
- 304 stainless steel construction with a polished No. 4 finish
Performance
- Knee space panels provide a finished look to the cabinetry, while providing structure and support
- This knee space panel is meant for use with an apron rail.
Dimensions
- LCKSH-ADJS: 48 in. L x 1 in. W x 30.6 in. H
Specifications
Specifications
| Product Type | Panel End Support |