New England Biolab Ltd Klenow Fragment (3'-5' exo-) 1000 units
Numéro de catalogue. 50811627
Klenow Fragment (3' to 5' exo-) is an N-terminal truncation of DNA Polymerase I which retains polymerase activity, but has lost the 5' to 3' exonuclease activity and has mutations (D355A, E357A) which abolish the 3' to 5' exonuclease activity.
Numéro de catalogue.
50811627
Fournisseur
New England Biolab LtdCode fournisseur
M0212M
Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.
Klenow Fragment (3' to 5' exo-) is an N-terminal truncation of DNA Polymerase I which retains polymerase activity, but has lost the 5' to 3' exonuclease activity and has mutations (D355A, E357A) which abolish the 3' to 5' exonuclease activity.