Learn More
KleenGuard™ Nemesis™ Safety Glasses
GREENER_CHOICE
Catalog No. 19005064
Change view
Click to view available options
Frame Color:
Black
Lens Tint:
Brown
Smoke
Lens Coating:
Anti-fog, Impact-resistant, UV-treated
Impact-resistant, UV-treated
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Frame Color Lens Tint Lens Coating
19005064 Black Brown Impact-resistant, UV-treated
19005075 Black Smoke Anti-fog, Impact-resistant, UV-treated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 19005064 Supplier KleenGuard™ Supplier No. 19642
KleenGuard™ Nemesis™ Safety Glasses feature durable lightweight construction for protection at work and at home.
  • Sporty, flexible, lightweight design
  • Optional Anti-Fog coating prevents fogging; ideal for humid environments and temperature variances
  • Meets ANSI Z87.1+ standards for personal eye protection
  • Polycarbonate lenses provide 99.9% UVA/ UVB/UVC protection
  • Patented FLEX-DRY™ nosepiece is designed to be flexible, to channel sweat away from the eyes, and to help prevent slippage down the nose
  • Recyclable via The RightCycle Program
  • Lens options include:
    • Smoke tint lenses with outer blue mirror finish coating is ideal for outdoor use where sunlight can cause eyestrain and fatigue
    • Copper blue shield lenses have a light brown tint and are designed to filter out blue light for enhanced sharpness and depth perception; ideal for low-light conditions
    • Bronze lenses provide visual definition with increased contrast and enhanced contours in most light conditions
    • Amber/yellow lenses block out blue light, making the environment appear brighter; ideal for low-light conditions and indoor use
    • IRUV Shade 3 lenses protect against infrared (IR) and ultraviolet radiation (UVA/UVB) during tasks like light cutting and brazing
    • IRUV Shade 5 lenses protect against infrared (IR) and ultraviolet radiation (UVA/UVB) during tasks like medium cutting brazing
    • Brown polarized lenses are effective for filtering scattered blue light to improve depth perception and contrast; ideal for outdoor use
Specifications
Lens Coating Impact-resistant, UV-treated
Certifications/Compliance ANSI Z87.1+
Lens Tint Brown
Product Type Safety Glasses
For Use With (Application) General-purpose eye protection
Frame Color Black
No. per Case 12 Pairs/Cs.
Material Plastic
Disposable Reusable
Lens Material Polycarbonate
Frame Material Plastic
UV Protection UV400
Show More Show Less