missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ansell KleenGuard™ A80 Chemical Permeation and Jet Liquid Protection Coveralls
Description
Level B/C suit made from durable fabric with exterior laminated with Saranex™ 23-P film. Suitable for numerous applications including chemical handling/mixing/clean-up, hazardous material handling, hazardous waste cleanup, industrial Hazmat teams, petrochemical, and utilities.
- Zipper front with taped storm flap
- Interior has comfortable, cloth-like feel against the skin
- Reflex™ Design provides 12% more chest room and 6% longer body length
Specifications
Specifications
| Product Type | Chemical Protective Clothing |
| Size | 4X-Large |
| Gender | Unisex |
| Material | Saranex 23-P Fabric |
| Color | White |
| Disposable | Single-use |
| Ankle Style | Attached Boots |
| Cuff Style | Elastic |
| Neck Style | Hooded |
| Hood Type | Attached |
| Show More |