missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ansell KleenGuard™ A80 Chemical Permeation and Jet Liquid Protection Coveralls
For chemical permeation and jet liquid protection
Supplier: Ansell 45645
Description
Level B/C suit made from durable fabric with exterior laminated with Saranex™ 23-P film. Suitable for numerous applications including chemical handling/mixing/clean-up, hazardous material handling, hazardous waste cleanup, industrial Hazmat teams, petrochemical, and utilities.
- Zipper front with taped storm flap
- Interior has comfortable, cloth-like feel against the skin
- Reflex™ Design provides 12% more chest room and 6% longer body length
Specifications
| 2X-Large | |
| White | |
| Elastic | |
| Elastic | |
| Taped | |
| Hooded (respirator-fit), zipper front with storm flap, elastic wrist and ankle, exterior lamination; Saranex™ 23-P Film Laminated to Durable Fabric | |
| 1.31 lb. |
| Saranex 23-P Fabric | |
| Elastic | |
| Hooded | |
| Front Zipper with Storm Flap | |
| Chemicals | |
| Chemical handling or mixing or cleanup, hazardous material handling, hazardous waste cleanup, industrial hazmat teams, petrochemicals, utilities and for chemical permeation and jet liquid protection | |
| 0.59 kg |