missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ansell KleenGuard™ A70 Chemical Spray Protection Apparel
Description
Use for hazardous waste remediation, environmental cleanup, petrochemical refining, acid/caustic handling, biohazard cleanup, tank cleaning, pharmaceutical manufacturing
- Material:1.5 mil polyethylene-film-coated spunbond polypropylene fabric
- Seamless chest area — primary splash zone
- Wick-away lining for enhanced comfort
- Bound seams
- Style: Hooded and booted, Zipper front, Taped storm flap, Elastic wrists
- Size: 5X-Large
- Color: Yelow
Order Info
Only available in North America
Compliance
NFPA 99 compliant antistat material
Specifications
Specifications
| Product Type | Chemical Protective Clothing |
| Size | 5X-Large |
| Gender | Unisex |
| Material | Spunbond Polypropylene |
| Color | Yellow |
| Disposable | Single-use |
| Ankle Style | Elastic, Attached Boots |
| Cuff Style | Elastic |
| Neck Style | Hooded |
| Hood Type | Elastic |
| Show More |