missing translation for 'onlineSavingsMsg'
Learn More

Ansell KleenGuard™ KGA300 Coveralls, Elastic Wrists, Ankles, No Hood
GREENER_CHOICE

Catalog No. p-7227816
Change view
Click to view available options
Size:
5X-Large/6X-Large
Small
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
17004474 Small
17004475 Medium
17004476 Large
17004477 X-Large
17004478 2X-Large
17004479 3X-Large
17004480 4X-Large
17004481 5X-Large/6X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Catalog No. 17004474 Supplier Ansell Supplier No. 41491
Only null left

KGA300 coveralls for surface preparation and paint protection are designed to protect against hazardous particulates and certain light liquid splashes while providing a comfortable fit and freedom of movement.

  • KGA300 liquid and surface prep and paint protection coveralls help protect against hazardous particulates and certain light liquid splashes
  • Made of disposable, lightweight microporous film laminate
  • Pass NFPA 99 for antistatic materials
  • Feature serged seams and a water resistant front zipper, and come in a variety of sizes and configurations
  • Human-centric Reflex™ design offers comfortable fit and freedom of movement
  • Eligible for recycling via RightCycle™ when used in non-hazardous applications as classified by federal, state, and local regulations
  • Zipper front, elastic wrists, ankles, and hood
  • Hood is designed to fit with respirator
  • Silicone- and latex-free

Specifications

Product Type General Purpose Coveralls
Size Small
Gender Unisex
Material Microporous Film Laminate
Color White
Disposable Single-use
Ankle Style Elastic
Cuff Style Elastic
Neck Style Collared
Hood Type Unhooded
Closure Type Front Zipper
Seam Type Serged
Sterility Non-sterile
Resistance Anti-static, Liquid, Particle
Certifications/Compliance NFPA 99
Latex Free Yes
Model KleenGuard™ KGA300
For Use With (Application) Surface Preparation and Painting
Quantity 25/Cs.
Show More Show Less