missing translation for 'onlineSavingsMsg'
Learn More

Ansell KleenGuard™ A40 Skid-Resistant Shoe Covers
GREENER_CHOICE

Catalog No. p-8874006
Change view
Click to view available options
Size:
Medium/Large
Small
X-Large/2X-Large
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19005119 Small
19005120 Medium/Large
19005121 X-Large/2X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 19005119 Supplier Ansell Supplier No. 51136
Only null left

Helps protect against slips, trips, and falls in a variety of work environments.

  • 7 in. high
  • Made from durable, gray, skid-resistant material
  • Offers up to 74% better traction on wet and dry surfaces than the leading competitor
  • Passes NFPA 99 for antistatic materials
  • Tested to withstand isocyanates and chromate dust

Specifications

Size Small
Material SMS
Color Gray
Disposable Single-use
Sterility Non-sterile
Closure Type Elastic Ankle
Sole Type Skid-resistant
Model 51136
For Use With (Application) Painting, Liquid Handling, General Manufacturing, Maintenance
Packaging Quantity 300/Cs.
Height (Metric) 17.78 cm
Product Type Shoe Covers
Show More Show Less