missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ansell KleenGuard™ A40 Reflex™ Liquid and Particle Protection Coveralls
Coveralls feature a respirator fit hood, zipper front, elastic wrists and ankles. Made of breathable, microporous film to keep out debris, non-hazardous liquid chemicals, and dry particulates.
Supplier: Ansell 47998
Description
- Feature respirator fit hood, zipper front, elastic wrists and ankles
- Made of breathable, microporous film to keep out debris, non-hazardous liquid chemicals and dry particulates
- Pass NFPA 99 for antistatic materials
- Pass ASTM F1670 testing for penetration of blood and bodily fluids
- Reflex sizing for a more comfortable fit and less rip outs
- Popular industries using this product include aviation, pharmaceutical manufacturing, liquid handling, general manufacturing
Specifications
| General Purpose Coveralls | |
| Unisex | |
| White | |
| Elastic | |
| Hooded | |
| Front Zipper | |
| Non-sterile | |
| NFPA 99, ASTM F1670 | |
| None |
| 2X-Large | |
| Microporous Film Laminate | |
| Limited-use | |
| Elastic | |
| Elastic, Respirator Fit | |
| Serged | |
| Liquid, Particle, Anti-static, Chemicals, Bloodborne Pathogens | |
| KleenGuard™ A40 Reflex™ | |
| Aviation, Pharmaceutical Manufacturing, General Manufacturing, Liquid Handling |