missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ansell KleenGuard™ A40 Coveralls, Elastic Wrists and Ankles
Protective coveralls are made of breathable, microporous film to keep out debris, non-hazardous liquid chemicals, and dry particulates.
Supplier: Ansell 44313
Description
Protective coveralls are made of breathable, microporous film to keep out debris, non-hazardous liquid chemicals, and dry particulates.
- Breathable, microporous film laminate fabric.
- Barrier protection against hazardous particulates and aerosols and against low-hazard liquid splash.
- ASTM F1670 for blood penetration and ASTM F1671 for bloodborne pathogens.
- ASTM F903 Penetration Testing provides passing results for isocyanate paint (40%) and Potassium Chromate (20%).
- Anti-stat according to NFPA 99.
- Features a zippered front and come in a variety of sizes.
- Eligible for recycling via RightCycle when used in non-hazardous applications as classified by federal, state, and local regulations.
- Elastic wrists and ankles.
- Human-centric Reflex™ design offers comfortable fit and freedom of movement.
Compliance
Meets ANSI/ISEA 101-1996; ASTM F1670 testing for penetration of blood and bodily fluids; NFPA 99 healthcare facilities criteria for antistatic materials.
Specifications
| General Purpose Coveralls | |
| Unisex | |
| White | |
| Elastic | |
| Collared | |
| Front Zipper | |
| Particle, Liquid | |
| KleenGuard™ A40 | |
| Elastic Wrists and Ankles |
| Large | |
| Microporous Film Laminate | |
| Limited-use | |
| Elastic | |
| Unhooded | |
| Non-sterile | |
| ASTM F1670, ASTM F1671, ASTM F903, NFPA 99 | |
| Fiberglass Manufacturing, Liquid Handling, Utilities, Pressure Washing, Tank Cleaning, Maintenance and General Manufacturing |