Learn More
Ansell KleenGuard™ A40 Coveralls, Elastic Wrists and Ankles
GREENER_CHOICE
Catalog No. 191202399H
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
5X-Large
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
191202399H 5X-Large
17000254 Small
191202399B Medium
191202399C Large
191202399D X-Large
191202399E 2X-Large
191202399F 3X-Large
191202399G 4X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Catalog No. 191202399H Supplier Ansell Supplier No. 30904
Only null left
Protective coveralls are made of breathable, microporous film to keep out debris, non-hazardous liquid chemicals, and dry particulates.
  • Breathable, microporous film laminate fabric.
  • Barrier protection against hazardous particulates and aerosols and against low-hazard liquid splash.
  • ASTM F1670 for blood penetration and ASTM F1671 for bloodborne pathogens.
  • ASTM F903 Penetration Testing provides passing results for isocyanate paint (40%) and Potassium Chromate (20%).
  • Anti-stat according to NFPA 99.
  • Features a zippered front and come in a variety of sizes.
  • Eligible for recycling via RightCycle when used in non-hazardous applications as classified by federal, state, and local regulations.
  • Elastic wrists and ankles.
  • Human-centric Reflex™ design offers comfortable fit and freedom of movement.
Specifications
Product Type General Purpose Coveralls
Size 5X-Large
Gender Unisex
Material Microporous Film Laminate
Color White
Disposable Limited-use
Ankle Style Elastic
Cuff Style Elastic
Neck Style Collared
Hood Type Unhooded
Closure Type Front Zipper
Seam Type Serged
Sterility Non-sterile
Resistance Particle, Liquid
Certifications/Compliance ASTM F1670, ASTM F1671, ASTM F903, NFPA 99
Model KleenGuard™ A40
No. of Pockets None
For Use With (Application) Fiberglass Manufacturing, Liquid Handling, Utilities, Pressure Washing, Tank Cleaning, Maintenance and General Manufacturing
Quantity 25/Cs.
Style Elastic Wrists and Ankles
Show More Show Less