Learn More
Ansell KleenGuard™ A40 Coveralls, Elastic Wrists and Ankles
Protective coveralls are made of breathable, microporous film to keep out debris, non-hazardous liquid chemicals, and dry particulates.
Supplier:  Ansell 30905
 View more versions of this product
Catalog No. 17000254
Only null left
Explore available promotions
Description
Description
Protective coveralls are made of breathable, microporous film to keep out debris, non-hazardous liquid chemicals, and dry particulates.
  • Breathable, microporous film laminate fabric.
  • Barrier protection against hazardous particulates and aerosols and against low-hazard liquid splash.
  • ASTM F1670 for blood penetration and ASTM F1671 for bloodborne pathogens.
  • ASTM F903 Penetration Testing provides passing results for isocyanate paint (40%) and Potassium Chromate (20%).
  • Anti-stat according to NFPA 99.
  • Features a zippered front and come in a variety of sizes.
  • Eligible for recycling via RightCycle when used in non-hazardous applications as classified by federal, state, and local regulations.
  • Elastic wrists and ankles.
  • Human-centric Reflex™ design offers comfortable fit and freedom of movement.
Compliance
Meets ANSI/ISEA 101-1996; ASTM F1670 testing for penetration of blood and bodily fluids; NFPA 99 healthcare facilities criteria for antistatic materials.
Specifications
Specifications
General Purpose Coveralls
Unisex
White
Elastic
Collared
Front Zipper
Particle, Liquid
KleenGuard™ A40
Elastic Wrists and Ankles
Small
Microporous Film Laminate
Limited-use
Elastic
Unhooded
Non-sterile
ASTM F1670, ASTM F1671, ASTM F903, NFPA 99
Fiberglass Manufacturing, Liquid Handling, Utilities, Pressure Washing, Tank Cleaning, Maintenance and General Manufacturing
Product Suggestions
Product Suggestions