missing translation for 'onlineSavingsMsg'
Learn More

Ansell KleenGuard™ A40 Liquid and Particle Protection Boot Covers
GREENER_CHOICE

Catalog No. p-200009434
Change view
Click to view available options
Size:
Medium/Large
Universal
X-Large/2X-Large
Height (English):
17 in.
18 in.
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size Height (English)
191202412 Universal 17 in.
19057521 Medium/Large 18 in.
19121718 X-Large/2X-Large 18 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 191202412 Supplier Ansell Supplier No. 44491
Only null left

A40 boot covers feature a highly durable PVC sole and are made with breathable, microporous film to keep out debris, non-hazardous liquid chemicals, and dry particulates.

  • Boot Covers available in 3 sizes, choose from 17 in. or 18 in. height
  • Elastic opening at top and elastic ankle
  • Serged seams
  • Highly durable PVC sole
  • Made to keep out debris, dry particulates, and liquid splashes
  • Adheres to NFPA 99 criteria for antistatic materials

Compliance

Meets NFPA™ 99 Health Care Facilities criteria for antistatic materials.

Specifications

Size Universal
Material Microporous Fabric
Coating Microporous Film Laminate
Color White
Sterility Non-sterile
Closure Type Elastic Top and Ankles
Sole Type PVC
Model 44491
Seam Type Serged
For Use With (Application) Aviation, Pharmaceutical Manufacturing, Liquid Handling, General Manufacturing
Certifications/Compliance NFPA 99, ASTM 1670
Packaging Quantity 400/Cs.
Height (English) 17 in.
Height (Metric) 43.1 cm
Resistance Fluid
Product Type Boot Covers
Show More Show Less