missing translation for 'onlineSavingsMsg'
Learn More

Ansell KleenGuard™ A30 Coveralls with Elastic Back, Wrists, Ankles, No Hood
GREENER_CHOICE

Catalog No. 19170387F
Change view
Click to view available options
Size:
Medium
Large
X-Large
2X-Large
4X-Large
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19170387F 4X-Large
296381594 Large
296381596 2X-Large
19170387A Medium
296381595 X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 19170387F Supplier Ansell Supplier No. 46107

Limited-use coveralls designed to protect against hazardous dry particles, aerosols, and low-hazardous limited liquid splash.

  • Air permeable SMS fabric incorporates a web of microfibers that filter particulates and water-based liquids
  • Suitable for many applications including pharmaceutical research and food processing
  • Provides protection from low-hazardous particles and liquids
  • Strong and abrasion-resistant
  • Suitable for multiple wearings
  • Seamless front to provide more protection in the primary exposure areas
  • Zipper front with 1 in. (2.5 cm) flap
  • Elastic back, wrists, ankles, no hood

Specifications

Product Type General Purpose Coveralls
Size 4X-Large
Gender Unisex
Material SMS Fabric
Color White
Disposable Limited-use
Ankle Style Elastic
Cuff Style Elastic
Neck Style Collared
Hood Type Unhooded
Closure Type Front Zipper
Sterility Non-sterile
Resistance Particle, Liquid
Back Style Elastic
Certifications/Compliance NFPA 99
Model KleenGuard™ A30
For Use With (Application) Pharmaceutical Research, Aerospace, Maintenance, General Manufacturing, Food
Show More Show Less