missing translation for 'onlineSavingsMsg'
Learn More

Ansell KleenGuard™ A20 Breathable Particle Protection Lab Coats

Catalog No. 19167145
Change view
Click to view available options
Closure Type:
Hook and Loop
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Closure Type
19167145 Hook and Loop
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19167145 Supplier Ansell Supplier No. 37427

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

KleenGuard™ A20 Lab Coats protect against hazardous and non-hazardous particulates

  • Designed to help protect against dirt, grime and dust
  • Made with breathable Microforce barrier fabric
  • Five layers of stacked barrier protection against hazardous particles
  • Serged seams
  • 4 hook and loop front closures
  • Elastic wrists
  • Knee-length; no pockets
  • Passes NFPA 99 for antistatic materials
  • Recyclable via The RightCycle Program

Specifications

Product Type General Purpose Lab Coat
Size 6X-Large
Gender Unisex
Material SMS
Color White
Closure Type Hook and Loop
Disposable Reusable
Sterility Non-sterile
No. of Pockets None
Certifications/Compliance TAA Compliant
For Use With (Application) Laboratories, Heavy Dust Environments
Show More Show Less