missing translation for 'onlineSavingsMsg'
Learn More

Kimberly-Clark Professional™ Kleenex™ Foam Skin Cleanser with Moisturizer

Catalog No. 17986055
Change view
Click to view available options
Volume (Metric):
1.2 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric)
17986055 1.2 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 17986055 Supplier Kimberly-Clark Professional™ Supplier No. 91592

Luxurious, moisturizing, pink foam soap.

  • Sealed container keeps soap free of contaminants
  • High capacity for less refilling, maintenance time
  • Easy-to-load cassette clicks right into place
  • Bottle is recyclable after pump is removed

Specifications

Product Type Disinfectant Soap
Color Pink
Form Foam
For Use With (Application) Disinfection
For Use With (Equipment) Kimberly-Clark Professional Cassette Skin Care Dispenser
Volume (Metric) 1.2 L