Learn More
Waters Limited Kit, I-Class PEEK Needle 10L
Catalog No. NC1530117
Encompass
Replacement 10ul PEEK needle assembly. Used in ACQUITY I-Class Fixed Loop (FL) Sample Managers.
Catalog No. NC1530117 Supplier Waters Limited Supplier No. 700005923
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Replacement 10ul PEEK needle assembly. Used in ACQUITY I-Class Fixed Loop (FL) Sample Managers.