Learn More
Thermo Scientific™ KingFisher™ SpeciTRAX™ Sample Transfer System, 1-year AB Assurance
The Thermo Scientific™ KingFisher™ SpeciTRAX™ Sample Transfer Instrument automates accessioning and sample transfer while minimizing the manual work of tube uncapping, barcode scanning, and pipetting.
Supplier:  Thermo Scientific™ A54552
Encompass
Catalog No. A54552
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description
Description
  • Instrument automates accessioning and sample transfer while minimizing the manual work of tube uncapping, barcode scanning, and pipetting
  • System combines new sample-handling automation technologies to offer ease of use, as well as increased throughput and safety, while delivering consistent, accurate results
  • Transfer 192 samples from capped sample tubes to two 96-deep-well plates in 40 minutes, all within a closed-door system
  • Tube decapping/recapping, barcode scanning, sample transfer
  • Precision pipetting done with fluid sensing tips
  • HEPA filtration and UV de-contamination
Specifications
Specifications
1 package
1 year manufacturer warranty plus 1 year service contract coverage
Product Suggestions
Product Suggestions