missing translation for 'onlineSavingsMsg'
Learn More

Kinetin, 99%

Catalog No. p-4510263
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC226500010 1 g Glass bottle
AC226500050 5 g Glass bottle
AC226500250 25 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC226500010 Supplier Thermo Scientific Chemicals Supplier No. 226500010
Only null left

CAS: 525-79-1 | C10H9N5O | 215.21 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 525-79-1
Molecular Formula C10H9N5O
Molecular Weight (g/mol) 215.21
InChI Key QANMHLXAZMSUEX-UHFFFAOYSA-N
Synonym kinetin, 6-furfurylaminopurine, 6-furfurylamino purine, 6-furfuryladenine, n6-furfuryladenine, n-furfuryladenine, cytokinin, n-furan-2-ylmethyl-9h-purin-6-amine, n6-furfurylamino purine, adenine, n-furfuryl
PubChem CID 3830
ChEBI CHEBI:27407
IUPAC Name N-(furan-2-ylmethyl)-7H-purin-6-amine
SMILES C1=COC(=C1)CNC2=NC=NC3=C2NC=N3

Specifications

Melting Point 269.0°C to 271.0°C
Color White to Beige
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Quantity 1 g
Beilstein 26,IV,3586
Merck Index 15,5359
Solubility Information Solubility in water: soluble. Other solubilities: freely soluble in dilute aquous hcl or naoh,slightly soluble in methanol and ethanol
Formula Weight 215.21
Percent Purity 99%
Physical Form Fine Crystalline Powder
Chemical Name or Material Kinetin
Show More Show Less
Hazard Category Warning
EINECSNumber 208-382-2

RUO – Research Use Only