Learn More
Kinematica Polytron™ PT-DA 1607/2EC Dispersing Aggregates
Supplier:  Kinematica 11030022
Catalog No. 08-451-171
Only null left
Explore available promotions
Description
Description
Specifications
Specifications
7.8 m/s
0.01 to 0.3 oz.
0.3 to 10mL
Dispersing of smallest amounts of plant, soft animal or soft human tissues in liquid for RNA extraction using test tubes and Eppendorf tubes
Product Suggestions
Product Suggestions