Learn More
Kinematica Polytron™ PT 10-35 GT & PT 3100D Homogenizer: Adapter
Catalog No. 13874619
Change view
Click to view available options
Packaging Quantity:
1 Ea.
Catalog No. Packaging Quantity
13874619 1 Ea.
1 options
Catalog No. 13874619 Supplier Kinematica Supplier No. 11035001
Accessory allows old model generators used on old model PT 3100 and PT 10-35 to be used with new models PT 3100D & PT 10-35 GT Homogenizer.
Users of older model Polytron™ Homogenizers (3100 & 10-35) can use their existing generators when using new models with the advanced quick connect feature.
Specifications
Product Type Adapter
Voltage 100/115/220/240 V
For Use With (Application) Ideal for dispersing aggregate with A coupling
For Use With (Equipment) Polytron™ PT 3100/1035 generator
Packaging Quantity 1 Ea.