missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Kinase Tracer 314, 25 μM

Catalog No. PV6087
Encompass
Change view
Click to view available options
Quantity:
25 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PV6087 25 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PV6087 Supplier Invitrogen™ Supplier No. PV6087

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Alexa Fluor™ Kinase Tracer 315 is detected by addition of a Eu-labeled anti-tag antibody.

  • Excitation/Emission (nm): 340/615, 665
  • Label or Dye: Alexa Fluor™ 647
  • Target Enzyme: Kinases (Multiple)
  • Validated Kinases: PIK3CD, PIK3CG, FRAP1 (mTOR)
  • In the LanthaScreen™ Eu Kinase Binding Assay, binding of an Alexa Fluor™ conjugate or tracer such as Kinase Tracer 314 to a kinase is detected by addition of a Eu-labeled anti-tag antibody. Binding of the tracer & antibody to a kinase results in a high degree of FRET, whereas displacement of the tracer with a kinase inhibitor results in a loss of FRET. Unlike many kinase activity assays, this assay is a simple mix and read assay, with no development steps.

Specifications

Assay Kinase Binding Assay
Assay Entry Biochemical binding/kinetic
Content And Storage Store at -20°C.
Excitation/Emission 340/615, 665
For Use With (Application) TR-FRET
For Use With (Equipment) Microplate Reader
Format 384-well Plate
Label or Dye Alexa Fluor™ 647
Quantity 25 μL
Readout Kinetic
Shipping Condition Dry Ice
Target Entry PIK3CG, PIK3CD/PIK3R1, mTOR, FRAP1
Type Kinase Tracer
Validated Kinases PIK3CD, PIK3CG, FRAP1 (mTOR)
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.