missing translation for 'onlineSavingsMsg'
Learn More

Promega Kinase Selectivity Profiling System: CMGC-2 + ADP-Glo™ Assay

Catalog No. PRV6857
Encompass
Change view
Click to view available options
Quantity:
8 x 50 Reactions
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PRV6857 8 x 50 Reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PRV6857 Supplier Promega Supplier No. V6857

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Easy-to-use kits for performing kinase selectivity profiling that rely on the ADP-Glo™ Kinase Assay technology. Each system includes kinase and substrate pairs in an easy-to-use 8-tube strip format optimized for fast and simple kinase profiling reactions.

  • Selectivity Screening of Library Compounds for Effect on CMGC Kinases
  • Kinase and substrate pairs organized in an easy-to-use 8-tube strip format
  • Simple reaction assembly and automation-friendly protocol
  • Streamilines profiling of hits and leads for potency against families of related kinases

Specifications

Quantity 8 x 50 Reactions
Content And Storage Catalog No. V6856 Kinase Selectivity Profiling System: CMGC-2, which includes:
  • 2 CMGC-2 Kinase Strip
  • 2 CMGC-2 Substrate/Co-Factor Strip
  • 1.5mL 5X Reaction Buffer A
  • 25μL DTT, 0.1M
Catalog No. V6930 ADP-Glo™ Kinase Assay, which includes:
  • 2mL ADP-Glo™ Reagent
  • 4mL Kinase Detection Reagent
  • 500μL Ultra Pure ATP, 10mM
  • 500μL ADP, 10mM
Components stored at different temperatures.
Format 384-well Plate
For Use With (Application)
  • Selectivity screening of library compounds for effects on target kinases
  • Profile hits and leads for their potency against families of related kinases
Product Type Kinase Selectivity Profiling System