missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Promega Kinase Selectivity Profiling System: AGC-1 + ADP-Glo™ Assay
Description
- Selectivity Screening of Library Compounds for Effect on AGC Family Kinases
- Kinase and substrate pairs organized in an easy-to-use 8-tube strip format
- Simple reaction assembly and automation-friendly protocol
- Streamilines profiling of hits and leads for potency against families of related kinases
Specifications
Specifications
| Quantity | 8 x 50 Reactions |
| Content And Storage | Catalog No. V6858 Kinase Selectivity Profiling System: AGC-1, which includes:
|
| Format | 384-well Plate |
| For Use With (Application) |
|
| Product Type | Kinase Selectivity Profiling System |