missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Kinase Buffer T (5X)

Catalog No. PV5373
Encompass
Change view
Click to view available options
Quantity:
4 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PV5373 4 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PV5373 Supplier Thermo Scientific™ Supplier No. PV5373

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Kinase buffer that has been specially optimized for use with the Type III PI4 kinases including PI4KA and PI4KB.

Kinase buffer that has been specially optimized for use with the Type III PI4 kinases including PI4KA and PI4KB.

Specifications

Concentration 5X
For Use With (Equipment) Pharma and biopharma, target and lead identification and validation
Packaging 1 Each
Physical Form Liquid
Quantity 4 mL
Form Liquid

For Research Use Only. Not for use in diagnostic procedures.