missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Kinase Buffer R (5X)

Catalog No. PV5101
Encompass
Change view
Click to view available options
Quantity:
4 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PV5101 4 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PV5101 Supplier Thermo Scientific™ Supplier No. PV5101

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Kinase buffer that has been specially optimized for use with the Class I & II PI3 kinases.

Kinase buffer that has been specially optimized for use with the Class I & II PI3 kinases.

Specifications

Concentration 5X
For Use With (Equipment) Pharma and biopharma, protein biology, drug discovery and development, target and lead identification and validation, kinase biology, biochemical kinase assays, Lanthascreen™ kinase activity assays, Adapta™ Universal kinase assays, enzyme and
Physical Form Liquid
Quantity 4 mL
Form Liquid

For Research Use Only. Not for use in diagnostic procedures.