missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Kinase Buffer Q (5X)

Catalog No. PV5125
Encompass
Change view
Click to view available options
Quantity:
4 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PV5125 4 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PV5125 Supplier Thermo Scientific™ Supplier No. PV5125

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Kinase buffer that has been specially optimized for use with PIK3C3 (hVPS34).

Kinase buffer that has been specially optimized for use with PIK3C3 (hVPS34).

Specifications

Concentration 5X
For Use With (Equipment) Pharma and biopharma, protein biology, drug discovery and development, target and lead identification and validation, kinase biology, biochemical kinase assays, Lanthascreen™ kinase activity assays, Adapta™ Universal kinase assays, enzyme and
Physical Form Liquid
Quantity 4 mL
Form Liquid

For Research Use Only. Not for use in diagnostic procedures.