missing translation for 'onlineSavingsMsg'
Learn More

Ansell Kimtech™ Pure M5 Pleat-Style Cleanroom Face Masks

Catalog No. 296360582
Change view
Click to view available options
Fastening Type:
Earloops
Ties
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Fastening Type
296360582 Ties
19062279 Earloops
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 296360582 Supplier Ansell Supplier No. 62742

Three-layer, pleat-style face masks for use in ISO Class 5 and higher cleanroom environments. Available with earloops or soft ties.

  • Three-layer design
  • Made with polyester and cellulose
  • Pleat-style masks available with earloops or soft ties
  • Fully enclosed bendable nosepiece
  • Popular for biotech, medical device, electronics and telecommunication environments
  • Ultrasonically bonded ties are durable
  • Double bagged and recommended for ISO Class 5 and higher cleanroom environments

Specifications

Product Type Cleanroom Face Mask
Color Blue
Fastening Type Ties
Material Polyester
Size Standard
Sterility Non-sterile
Style Pleated
Cleanroom Class Class 100 (ISO 5)
Construction 3-ply
Disposable Single-use
Filter Efficiency 99%
For Use With (Application) Biotech, Medical Devices, Electronics, Telecommunications
Model 62742
Packaging Type Double-bagged
Series M5
Show More Show Less