missing translation for 'onlineSavingsMsg'
Learn More

Ansell Kimtech™ A8 Cleanroom Coveralls
GREENER_CHOICE

Catalog No. 19169222E
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19169222E 2X-Large
19169222A Small
19169222B Medium
19169222C Large
19169222D X-Large
19169222F 3X-Large
19169222G 4X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. 19169222E Supplier Ansell Supplier No. 47955
Edge

Cleanroom coveralls are designed to enhance comfort and reduce heat stress in ISO 7 and 8 environments.

Kimtech™ A8 Cleanroom Coveralls are made with a lightweight, innovative fabric that moves with the worker, and feature built-in thumb loops to protect your people and processes, and a high neck collar to control contamination risk. These white coveralls also have elastic wrists, ankles and waist.

They are designed for protection against dust, chemicals and other contaminants and have a BFE of 94%. The white coveralls are designed for applications including ISO 7/8 cleanrooms, Grade C/D, non-sterile pharmaceutical manufacturing, bio-medical research, pharmaceutical compounding and biotechnology.

Plus, these protective suits are recyclable through the RightCycle Program.

  • 25 Kimtech™ A8 Coveralls/Case
  • Helps protect against dust, chemicals and other contaminants; BFE of 94%; ISO 7/8 cleanrooms
  • Low lint for contamination control
  • Recyclable through the RightCycle Program, which turns used lab coats into raw materials for eco-friendly consumer products

Specifications

Product Type Cleanroom Coveralls
Size 2X-Large
Material SMS
Color White
Disposable Single-use
Cuff Style Thumb Loop
Neck Style Collared
Sterility Non-sterile
Resistance Anti-static
Collar Type Standing
Certifications/Compliance PPE Cat III directive 89/686/EEC, Type 6 limited chemical splash protection; Type 5 particle protection
Model Kimtech™ A8
For Use With (Application) Pharmaceutical, Packing, Distribution, Production, Biotechnology
Packaging Double Bagged
Cleanroom Class Class 10,000-100,000 (ISO 7-8)
Show More Show Less