missing translation for 'onlineSavingsMsg'
Learn More

Ansell Kimtech Pure™ A5 Sterile Elasticized Hood
GREENER_CHOICE

Catalog No. 19169987
Change view
Click to view available options
Quantity:
100/Cs.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
19169987 100/Cs.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19169987 Supplier Ansell Supplier No. 40616

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Offers an uninterrupted seal and customized fit for anyone who wears a full-face respirator.

  • Stretch fit elastic hood and opening
  • Works with many respirator styles and brands
  • Tunneled over seams to prevent particle shedding
  • Clean-Don ties for easy donning and a more secure fit
  • Great for small faces
  • Reduces risk of contamination

Specifications

Size Universal
Material Polypropylene SMS Fabric
Color White
Closure Type Ties
Disposable Single-use
Model 40616
Sterility Sterile
For Use With (Equipment) Full face respirators
Quantity 100/Cs.
Hood Type Elasticized
Cleanroom Class Class 100 (ISO 5)
Show More Show Less