missing translation for 'onlineSavingsMsg'
Learn More

Ansell Kimtech™ Polaris™ Xtra Nitrile Exam Gloves
GREENER_CHOICE

Catalog No. 17103191E
Change view
Click to view available options
Size:
X-Small
Small
Medium
Large
X-Large
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
17103191E X-Large
17103191A X-Small
17103191B Small
17103191C Medium
17103191D Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 17103191E Supplier Ansell Supplier No. 62764

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Kimtech™ is your partner in driving progress. Tested against some of the harshest chemicals used in your work environment, Kimtech™ Polaris™ Exam Nitrile Gloves meet and exceed the industry standards for exam gloves.

  • The proprietary high-quality nitrile formulation delivers the highest level of protection in the Kimtech™ exam glove portfolio and has been tested for use with chemotherapy drugs and fentanyl.
  • With an average tensile strength that is more than double the industry standard, Kimtech™ Polaris™ provides the strength and comfort needed for performance and productivity; it's even certified by US Ergonomics.
  • Eligible for recycling through The RightCycle™ Program, 100% of your non-hazardous Kimtech™ Polaris™ Nitrile Exam Gloves can be diverted from the landfill.
  • With protection, durability, and comfort you can count on, these gloves are built for science, made for you
  • Ambidextrous

Specifications

Chlorinated No
Accelerator Free No
Applications Laboratory
AQL 0.65
Color Dark Magenta
Disposable Single-use
Exterior Finish Fingertip Textured
Finger Thickness (English) 7.48 mil
Finger Thickness (Metric) 0.19 mm
Latex Free Yes
Material Nitrile
Palm Thickness (English) 5.91 mil
Palm Thickness (Metric) 0.15 mm
Powdered Powder Free
Product Type Nitrile Examination Glove
Size X-Large
Sterility Non-sterile
Thickness (English) 5.91 mil
Thickness (Metric) 0.15 mm
FDA 510 (k) Yes
Glove Cuff Style Beaded
Length (English) 12 in.
Length (Metric) 30.48 cm
Show More Show Less