missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ansell Kimtech™ Polaris™ Xtra Nitrile Exam Gloves

Description
- The proprietary high-quality nitrile formulation delivers the highest level of protection in the Kimtech™ exam glove portfolio and has been tested for use with chemotherapy drugs and fentanyl.
- With an average tensile strength that is more than double the industry standard, Kimtech™ Polaris™ provides the strength and comfort needed for performance and productivity; it's even certified by US Ergonomics.
- Eligible for recycling through The RightCycle™ Program, 100% of your non-hazardous Kimtech™ Polaris™ Nitrile Exam Gloves can be diverted from the landfill.
- With protection, durability, and comfort you can count on, these gloves are built for science, made for you
- Ambidextrous
Specifications
Specifications
| Chlorinated | No |
| Accelerator Free | No |
| Applications | Laboratory |
| AQL | 0.65 |
| Color | Dark Magenta |
| Disposable | Single-use |
| Exterior Finish | Fingertip Textured |
| Finger Thickness (English) | 7.48 mil |
| Finger Thickness (Metric) | 0.19 mm |
| Latex Free | Yes |
| Show More |