missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ansell Kimtech™ Polaris™ Nitrile Exam Gloves
Kimtech™ is your partner in driving progress. Tested against some of the harshest chemicals used in your work environment, Kimtech™ Polaris™ Exam Nitrile Gloves meet and exceed the industry standards for exam gloves.
Supplier: Ansell 62773
Description
- The proprietary high-quality nitrile formulation delivers the highest level of protection in the Kimtech™ exam glove portfolio and has been tested for use with chemotherapy drugs and fentanyl.
- With an average tensile strength that is more than double the industry standard, Kimtech™ Polaris™ provides the strength and comfort needed for performance and productivity; it's even certified by US Ergonomics.
- Eligible for recycling through The RightCycle™ Program, 100% of your non-hazardous Kimtech™ Polaris™ Nitrile Exam Gloves can be diverted from the landfill.
- With protection, durability, and comfort you can count on, these gloves are built for science, made for you
- Ambidextrous
Specifications
| No | |
| Laboratory | |
| Dark Magenta | |
| Fingertip Textured | |
| 0.15 mm | |
| Nitrile | |
| 0.15 mm | |
| Nitrile Examination Glove | |
| Non-sterile | |
| 0.15 mm | |
| Beaded | |
| 24.13 cm |
| No | |
| 0.65 | |
| Single-use | |
| 5.91 mil | |
| Yes | |
| 5.91 mil | |
| Powder Free | |
| Large | |
| 5.91 mil | |
| Yes | |
| 9.5 in. |