missing translation for 'onlineSavingsMsg'
Learn More

Ansell Kimtech™ A7 Certified Liquid Barrier Gowns
GREENER_CHOICE

Catalog No. p-7078341
Change view
Click to view available options
Size:
Large/X-Large
Small/Medium
2X-Large
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
17986050 Small/Medium
17986051 Large/X-Large
17986052 2X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 17986050 Supplier Ansell Supplier No. 47992
Only null left

Non-sterile gowns deliver stringent barrier protection and provide contamination control for clinical, chemotherapy, and laboratory settings.

  • Liquid barrier gowns
  • Seamless front design
  • Easy to don-and-doff design to reduce contamination risk
  • Material offers enhanced liquid barrier protection
  • Fabric tested for blood and blood-borne pathogen penetration, chemotherapy drug and hormone permeation
  • Passes ASTM F1670 and ASTM F1671
  • Built-in thumb loops help maintain glove and garment interface
  • Low lint to minimize contamination
  • Meets USP 800 guidelines
  • Bulk-packaged and available in a range of sizes

Compliance

Meets USP 800 guidelines

Specifications

Size Small/Medium
For Use With (Application) Clinical, Pharmaceutical, Laboratories, Chemotherapy
Gender Unisex
Product Type Controlled Environments Gown
No. of Pockets None
Packaging Type 10 Bags of 10 Gowns
Material Polyethylene, Polypropylene
Color Blue
Disposable Single-use
Cuff Style Thumb Loop
Model 47922
Seam Type Seamless Front
Certifications/Compliance ASTM 1670, ASTM 1671, Meets USP 800 Guidelines
Autoclavable Not Autoclavable
Show More Show Less