Learn More
DWK Life Sciences Kimble™ White Urea Closures with PTFE-Faced Foam-Backed Rubber Liner
White closures with rubber liner
$129.38 - $1,682.64
Specifications
Color Black
Material Phenolic Resin
View More Specs
Products 5
Catalog Number Mfr. No. Neck Finish Price Quantity & Availability  
Catalog Number Mfr. No. Neck Finish Price Quantity & Availability  
03340123B
View Documents Promotion Details
DWK Life Sciences
73802U13425
13-425
Pack of 144 Each for $129.38
Case of 3 Pack for $388.14
Only null left
03340123A
View Documents Promotion Details
DWK Life Sciences
73802U8425
8-425
Case of 432 Each for $549.22
Only null left
03340123C
View Documents Promotion Details
DWK Life Sciences
73802U15425
15-425
Case of 432 Each for $567.53
Only null left
03340123D
View Documents Promotion Details
DWK Life Sciences
73802U18400
18-400
Pack of 144 Each for $176.25
Case of 3 Pack for $528.75
Only null left
03340123F
View Documents Promotion Details
DWK Life Sciences
73802U24400
24-400
Case of 432 Each for $1,682.64
Only null left
Description
Description
White closures with rubber liner
  • White urea
  • PTFE-faced foam-backed rubber liner
Specifications
Specifications
Black
Phenolic Resin