Learn More
DWK Life Sciences Kimble™ ValueWare™ Narrow-Mouth Erlenmeyer Flasks
Economical and versatile; ideal for general laboratory usage.
$351.63 - $351.63
Specifications
| Closure Material | Rubber |
|---|---|
| Description | White Ceramic Enamel scale graduations; Top style: Reinforced and tooled with rounded finish beaded |
| Material | Borosilicate Glass |
| Mouth Style | Narrow |
Description
- Kimax™ KG-33 glass Reinforced tops tooled with a rounded finish — contain more glass for maximum mechanical strength
- Thick-walled body has long tapered outside contour to minimize chipping when struck or rubbed together
- Durable white ceramic enamel scales indicate approximate volumes at various levels — useful for measuring and mixing solutions where a high degree of accuracy is not necessary
Compliance
Glass meets Type 1, Class A, ASTM™ E-438 standards. Designed to ASTM Specification E1404, Type I, Class 1 specification requirements.
Specifications
| Rubber | |
| White Ceramic Enamel scale graduations; Top style: Reinforced and tooled with rounded finish beaded | |
| Borosilicate Glass | |
| Narrow |