missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DWK Life Sciences Kimble™ PTFE Flathead Stopper
Color button insert serves as a means of color-coding
$728.10 - $728.10
Specifications
| Material | PTFE |
|---|---|
| Shape | Flat Grip |
| For Use With (Application) | Used with joint designations |
| Catalog Number | Mfr. No. | Diameter (Metric) | Color | Stopper Number | Price | Quantity | |||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Diameter (Metric) | Color | Stopper Number | Price | Quantity | |||
|
14-642-17C
|
DWK Life Sciences
41941R24 |
2.4 cm | Blue | 24 |
Case of 6 Each for $728.10
|
|
|||
Description
- Bottle and Apparatus Style
- Kimble
- Stoppers can be used with joint designations
- Size 24
- Approx. Dia. at Large End: 24.0mm
- Used with Joint 24/40
Specifications
| PTFE | |
| Flat Grip | |
| Used with joint designations |