missing translation for 'onlineSavingsMsg'
Learn More

DWK Life Sciences Kimble™ Phenolic Caps with Tinfoil Liners

Catalog No. 13757154
Change view
Click to view available options
Neck Finish:
20-400
22-400
24-400
28-400
33-400
38-400
45-400
48-400
53-400
58-400
63-400
70-400
89-400
13 product options available for selection
Product selection table with 13 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Neck Finish
13757154 89-400
13757144 28-400
13757143 24-400
13757141 20-400
13757142 22-400
13757145 33-400
13757146 38-400
13757148 45-400
13757152 63-400
13757150 53-400
13757149 48-400
13757151 58-400
13757153 70-400
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 13 options available.
13 options
Not recommended for acids and alkalis.
Catalog No. 13757154 Supplier DWK Life Sciences Supplier No. 75203G89400
Not recommended for acids and alkalis.

Has good resistance to alcohols, hydrocarbons, ketones, oils; not recommended for acids and alkalis.

  • Economical general purpose phenolic screw cap with a tinfoil liner
  • Color: black
  • Best suited for environmental sampling applications
  • Foil is laminated to paper and bonded to a 0.035-in. pulpboard

Specifications

Color Black
Material Phenolic Resin
No. per Case 144 Cs.
Neck Finish 89-400
Product Type Phenolic Cap
For Use With (Equipment) Bottle
Closure Liner Tin