Learn More
DWK Life Sciences Kimble™ Phenolic Caps with Tinfoil Liners
Has good resistance to alcohols, hydrocarbons, ketones, oils; not recommended for acids and alkalis.
Supplier: DWK Life Sciences 75203G24400
Alert:
Not recommended for acids and alkalis.
Description
- Economical general purpose phenolic screw cap with a tinfoil liner
- Color: black
- Best suited for environmental sampling applications
- Foil is laminated to paper and bonded to a 0.035-in. pulpboard
Chemical Identifiers
| Black | |
| 144 Cs. | |
| Bottle |
| Phenolic Resin | |
| Phenolic Cap | |
| Tin |
Specifications
| Black | |
| 144 Cs. | |
| Phenolic Cap | |
| Tin |
| Phenolic Resin | |
| 24-400 | |
| Bottle |