missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DWK Life Sciences Kimble™ Linear HDPE Stoppers
Closed bottom end fits the standard taper neck
$20.19 - $162.33
Specifications
| Color | Yellow, Blue |
|---|---|
| Material | HDPE |
| Product Type | Tapered Stopper |
| For Use With (Application) | Used as replacement parts for items that are ground to standard taper stopper dimensions |
| Catalog Number | Mfr. No. | Size | Stopper Number | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Size | Stopper Number | Price | Quantity | ||||
|
14-644-1E
|
DWK Life Sciences
28160R19 |
19 | 19 |
Case of 6 Each for $126.57
|
|
||||
|
14-644-1G
|
DWK Life Sciences
28160R27 |
27 | 27 |
Case of 6 Each for $159.40
|
|
||||
|
14-644-1B
|
DWK Life Sciences
28160R9 |
9 | 9 |
Case of 6 Each for $92.19
|
|
||||
|
14-644-1C
|
DWK Life Sciences
28160R13 |
13 | 13 |
Case of 6 Each for $96.53
|
|
||||
|
14-644-1F
|
DWK Life Sciences
28160R22 |
22 | 22 |
Case of 6 Each for $162.33
|
|
||||
|
14-644-1D
|
DWK Life Sciences
28160R16 |
16 | 16 |
Each for $20.19
Case of 6 Each for $121.14
|
|
||||
Description
Closed bottom end fits the standard taper neck
- Heavy-duty Polyethylene stopper
- Flange on stopper protects glass
Specifications
| Yellow, Blue | |
| HDPE | |
| Tapered Stopper | |
| Used as replacement parts for items that are ground to standard taper stopper dimensions |